Propylester av gallussyre ≥98% (by HPLC) for histology and pathology, for diagnostic, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning


M-Clarity™ quality level = MQ100

02370-100G
02370-100GEA 775 NOK
02370-100G
Propylester av gallussyre ≥98% (by HPLC) for histology and pathology, for diagnostic, Sigma-Aldrich®
Propylester av gallussyre
Formel: C₁₀H₁₂O₅
Kokepunkt: 449 °C (1013 hPa)
Smeltepunkt: 145…148 °C
Tetthet: 1,21 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002196
CAS-nummer: 121-79-9
EINECS: 204-498-2
Merck Indeks: 12,08044

Bestill nå

Spesifikasjon Testresultater

Appearance (colour) White to faint brown
Appearance (form) Powder
Conforms to structure ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR