N-Hydroxy Desloratadine, Pharmaceutical Analytical Impurities

Leverandør: USP

1A01660
1A01660.EA 10775 NOK
1A01660.
N-Hydroxy Desloratadine, Pharmaceutical Analytical Impurities
Standarder Organiske standarder

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

Bestill nå

SPESIFIKASJONER

Beskrivelse N-Hydroxy Desloratadine
Environmentally Preferable
Synonym(s) 4-(8-Chloro-5,6-dihydro-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-piperidin-1-ol
CAS-nr 1193725-73-3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR