Homogeniser, T 50 digital ULTRA-TURRAX®
Leverandør: IKA
3787002 3787000 3787003 9024500
431-0181EA 27150 NOK
431-0181 431-0180 431-0182 IKAA9024500
Homogeniser, T 50 digital ULTRA-TURRAX®
Homogenisatorer Homogenisatorer, bordmodell
Stand model, volume range 250 to 30000 ml. Reproducible operation due to constant speed, even with changes in viscosities.
  • Digital display for precise monitoring of set and actual speeds
  • Wide selection of dispersing tools to suit your application
  • Motor protected against overload
  • Smooth start
  • Error code display
Optional shaft 431-2961 enables the T 50 basic to be used as a high-speed stirrer.
IP protection class according to DIN EN 60529: IP 20
Sertifiseringer: IP protection class according to DIN EN 60529: IP 20.
Tilbehørs informasjon: More Product Information