Incubating shaker, KS 3000 i control and KS 3000 ic control
Leverandør: IKA
3940100 3940102 3940103 3940003 3940000 3940002
444-0809EA 0 NOK
444-0809 444-0810 444-0811 444-0808 444-0799 444-0807
Incubating shaker, KS 3000 i control and KS 3000 ic control
Risteapparater Risteinkubatorer
Compact incubator shakers designed for safe, unattended operation in a temperature controlled environment. The units have a large LED display for speed, temperature and timer. An integrated PID temperature control allows the use of PT1000 temperature sensors to assure highly accurate temperature control. The electronically controlled incubators come equipped with RS232 and USB ports for PC interface. All functions can be controlled and documented using optional labworldsoft® software. In addition the  KS 3000 ic control has a built in cooling coil for connection to an external coolingMore Product Information