Anti-Gentamicin Rabbit polyclonal antibody

Leverandør: Biorbyt
ORB10384-100 ORB10384-200
BIRBORB10384-100EA 0 NOK
BIRBORB10384-100 BIRBORB10384-200
Anti-Gentamicin Rabbit polyclonal antibody
Antistoffer
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: Gentamicin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn Gentamicin
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Immunogen Synthetic peptide
Skjema liquid
Rensing purified by affinity chromatography
Konsentrasjon 0,5 mg/ml
Lagringsbuffer 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR