Anti-KLH Rabbit Polyclonal Antibody

Leverandør: Biorbyt
ORB10961-100 ORB10961-200
BIRBORB10961-100EA 0 NOK
BIRBORB10961-100 BIRBORB10961-200
Anti-KLH Rabbit Polyclonal Antibody
Antistoffer
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR