Vann for biotechnology nuclease-free, 0.2 µm filtered

Leverandør: VWR Chemicals


0,2 µm filtrert

E476-500MLEA 655 NOK
E476-500ML E476-1L E476-100ML
Vann for biotechnology nuclease-free, 0.2 µm filtered
Vann

Nuclease-free water treated with diethylpyrocarbonate (DEPC) for nucleic acid applications.

Formel: H₂O
Kokepunkt: 100 °C (1013 hPa)
Smeltepunkt: 0 °C
Tetthet: 1 g/cm³ (4 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011332
CAS-nummer: 7732-18-5
EINECS: 231-791-2
Merck Indeks: 13,10098

Bestill nå

Spesifikasjon Testresultater

DNase NONE
Protease NONE
RNase NONE
Sterility PASS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR