Soap dispensers, Stoko Vario® mat
Leverandør: Deb-STOKO
PN89745X01
EVONPN89745X01EA 0 NOK
EVONPN89745X01
Soap dispensers, Stoko Vario® mat
Såpedispensere og Lotion Såpedispensere
This dispenser made from steel is available with or without long arm lever.
  • Can be locked
  • Variable use of all 1000 and 2000 ml soft bottles
  • Easy to use and refill, rugged construction and reliable operation
Colour: White.