Volumetric flasks, class A, blue markings, BLAUBRAND®
Leverandør: Brand
2.37263 2.37264 2.37265 2.37266 2.37288 2.37289 2.37262 2.37294 2.37261 2.37260 2.37259 2.37258 2.37256 2.37257
BRND2.37263EA 1575 NOK
BRND2.37263 BRND2.37264 BRND2.37265 BRND2.37266 BRND2.37288 BRND2.37289 BRND2.37262 BRND2.37294 BRND2.37261 BRND2.37260 BRND2.37259 BRND2.37258 BRND2.37256 BRND2.37257
Volumetric flasks, class A, blue markings, BLAUBRAND®
Kolber Målekolber
Borosilicate glass 3.3, with glass stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
  • Total order from one batch, supplied with batch certificate
*Wide neck.
Sertifiseringer: DIN EN ISO 1042.
Bestillingsinformasjon: Supplied with a batch certificate. Volumetric flasks also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.