Fluorometric HTS assay kits, Caspase-3 and Caspase-8
Leverandør: Biotium
30009-2EA 3500 NOK
30009-2 30009-1 30009-3 30012-2 30012-1 30012-3
Fluorometric HTS assay kits, Caspase-3 and Caspase-8
Assay Enzymanalyser
Caspase-3 and Caspase-8 Fluorometric HTS Assay Kits provide a homogenous assay system for fast and highly sensitive detection of Caspase-3 or Caspase-8 activity by fluorescence in enzymatic reaction or mammalian cells.
  • Single-step homogenous assay specifically designed for HTS-based detection
  • Fast enzyme kinetics
  • Sensitive green fluorescent readout
Caspase-3 DEVD-R110 Fluorometric HTS Assay Kit includes the fluorogenic substrate (Ac-DEVD)2-R110, which contains two DEVD tetrapeptides and is completely hydrolysed by the enzyme in two successive steps. Cleavage of the first DEVD peptide results in the monopeptide Ac-DEVD-R110 intermediate, which has absorption and emission wavelengths similar to those of R110 (λEx/λEm = 496/520 nm) but has only about 10% of the fluorescence of the latter. Hydrolysis of the second DEVD peptide releases the dye R110, leading to a substantial fluorescence increase.
The Caspase-3 Fluorometric HTS Assay Kit includes Ac-DEVD-CHO, which is a caspase-3 inhibitor and can be used as a negative control. Also, R110 is provided in the kit for generating a standard curve, which can be used for quantifying caspase-3 activity. One mL cell lysis/assay buffer is sufficient for 10 assays in 96-well format.
Caspase-8 IETD-R110 Fluorometric HTS Assay Kit includes the fluorogenic substrate (Ac-IETD)2-R110, which contains two IETD tetrapeptides and is completely hydrolysed by the enzyme in two successive steps. Cleavage of the first IETD peptide results in the monopeptide Ac-IETD-R110 intermediate, which has absorption and emission wavelengths similar to those of R110 (λEx/λEm = 496/520 nm) but has only about 10% of the fluorescence of the latter. Hydrolysis of the second IETD peptide releases the dye R110, leading to a substantial fluorescence increase.
The Caspase-8 Fluorometric HTS Assay Kit includes Ac-IETD-CHO, which is a caspase-8 inhibitor and can be used as a negative control. Also, R110 is provided in the kit for generating a standard curve, which can be used for quantifying caspase-8 activity.
Note: While caspase-8 preferentially cleaves the consensus sequence IETD compared to other substrate sequences, other caspases such as caspase-3 also can cleave IETD efficiently. Overlapping caspase substrate recognition limits the usefulness of caspase substrate peptides for distinguishing between different caspase activities in cell lysates.