Syringe filters, prefilter, sterile, GD/X™, Whatman™
Leverandør: Whatman products (Cytiva)
6896-2502 6896-2504 6900-2502 6900-2504 6901-2502 6902-2504 6901-2504
514-0537EA 4775 NOK
514-0537 514-0538 514-0539 514-0540 514-0541 516-6280 514-0542
Syringe filters, prefilter, sterile, GD/X™, Whatman™
Filter Sprøytefilter
Cytiva's Whatman GD/X sterile syringe filter is designed to support filtration of viscous, hard-to-filter samples with high solids content.
  • Reduces need for multiple syringe filters by processing three to seven times more sample volume than syringe filters without prefilters
  • Female luer lock for leak-free connections
  • High-efficiency Whatman GMF 150 and GF/F pre-filter media
  • Eliminates or greatly reduces the need to carry out separate prefiltration step for difficult-to-filter samples
  • Recyclable after use
Cytiva's GD/X sterile syringe filter isMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Dimensjoner 21,6×29,8 mm
Maks. bemanningstrykk 5,2 bar
Vekt 3 g