Screen Quest™ Fluo-4 No Wash Calcium Assay Kit
Leverandør: AAT BIOQUEST
36325 36326
AATB36325EA 7200 NOK
AATB36325 AATB36326
Screen Quest™ Fluo-4 No Wash Calcium Assay Kit
Assay Cellulær analyse
Calcium flux assays are preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
  • Well-cited calcium probe
  • Contains all necessary components
  • No wash step required
  • Robust and adaptable protocol
  • Multiple research applications
NB: For research use only.