Antimon ≥99.99% (metals basis), ingot

Leverandør: Thermo Fisher Scientific
Warning

00030.A1
00030.A1EA 28025 NOK
00030.A1
Antimon ≥99.99% (metals basis), ingot
Antimony

Purity: 99.99% CAS No.: 7440-36-0 Odour: Odourless MDL No.: MFCD00134030

Formel: Sb
Kokepunkt: 1637 °C (1013 hPa)
Smeltepunkt: 630 °C
Tetthet: 6,684 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00134030
CAS-nummer: 7440-36-0
EINECS: 231-146-5
FN: 2871
ADR: 6.1,III
Merck Indeks: 12,00733

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR