VWR® Incubating Rocking Platform Shaker
Leverandør: VWR Collection
444-0762EA 41175 NOK
444-0762 444-0764 444-0763
VWR® Incubating Rocking Platform Shaker
Risteapparater Risteinkubatorer
The incubating rocking platform shaker combines smooth rocking motion and incubation in one compact bench top unit. The spill resistant design channels fluids away from internal components. The microprocessor control provides electronic tilt angle adjustment which allows the user to easily adjust the rocking angle from zero to 15° while unit is operating. The PID temperature controller maintains precise temperature control from ambient 5 to 65 °C.
  • Electronic tilt adjustment from zero to 15° while unit is operating
  • LED display shows temperature, speed, tilt