Ammoniumacetat ≥97%, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Warning

0011.0500EA 960 NOK
0011.0500 0011.1000 0011.9020BB
Ammoniumacetat ≥97%, BAKER ANALYZED® ACS, J.T.Baker®
Ammoniumacetat
Formel: H₃CCOONH₄
Smeltepunkt: 114 °C
Tetthet: 1,17 g/cm³ (20 °C)
Flash punkt: 136 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013066
CAS-nummer: 631-61-8
EINECS: 211-162-9
Merck Indeks: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Meets ACS Specifications
Assay min. 97 %
Insoluble Matter max. 0.005 %
Nitrate (NO3) max. 0.001 %
pH of 5% Solution at 25°C 6.7 - 7.3
Residue after Ignition max. 0.01 %
Sulfate (SO4) max. 0.001 %
Trace Impurities (in ppm) max. 5
Chloride (Cl) max. 5
Iron (Fe) max. 5
Heavy Metals (as Pb) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR