Etanolamin, Supelco®

Leverandør: Merck
Danger

Synonymer: 2-Aminoethanol, Monoethanolamine

MQ100

00278-5ML-F
00278-5ML-FEA 1550 NOK
00278-5ML-F
Etanolamin, Supelco®
Etanolamin
Formel: NH₂CH₂CH₂OH
MW: 61.08 g/mol
Kokepunkt: 171
Smeltepunkt: 10.5
Tetthet: 1.0164 g/cm³ (20 °C)
Flash punkt: 93
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008183
CAS-nummer: 141-43-5
EINECS: 205-483-3
FN: 2491
ADR: 8 III
Merck Indeks: 13,03762
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=388ecb2c206503a3db082d91353101a3bbae24de&vid=99ca3d2ea81643daf6bf23fbf436e432a3ae6dc1&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR