Acetonitril ≥99.9% (by GC), CHROMASOLV™, gradient grade suitable for PAH analysis, suitable for PAH analysis, Riedel-de Haën™

Leverandør: Honeywell Chemicals
Danger

Synonymer: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

00683-2.5L
00683-2.5LEA 5875 NOK
00683-2.5L
Acetonitril ≥99.9% (by GC), CHROMASOLV™, gradient grade suitable for PAH analysis, suitable for PAH analysis, Riedel-de Haën™
Acetonitril

  • Same high quality as CHROMASOLV HPLC
  • Additional testing for HPLC via gradient analysis and UV-VIS spectroscopy
  • Low UV profile by HPLC
  • Low impurity gradient LC baseline


Beilstein Number: 741857

Formel: H₃CCN
MW: 41.05 g/mol
Kokepunkt: 82 °C (1.013 hPa)
Smeltepunkt: –46 °C
Tetthet: 0.7857 g/cm³ (20 °C)
Flash punkt: 6 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001878
CAS-nummer: 75-05-8
EINECS: 200-835-2
FN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Indeks: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (GC) Min. 99.9 %
Density (D 20/4) 0.765 - 0.794
Refractive index (n 20/D) 1.343 - 1.345
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.02 %
Free acid (as CH₃COOH) Max. 0.005 %
Free alkali (as NH₃) max. 0.0005 %
Suitability test (HPLC) complying
Absorbance at 230 nm max. 0.005
Absorbance at 215 nm max. 0.01
Absorbance at 200 nm max. 0.04
Fluorescent substances complying
Refractive Index n20/D 1.344 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR