VWR® Q Path® Formaldehyde Neutraliser
Leverandør: VWR Chemicals
00699030.EA 1550 NOK
00699030.
VWR® Q Path® Formaldehyde Neutraliser
Rengjøringsløsninger Detergenter og overflatevasking
Acidic neutraliser and pre-cleaner based on organic acid.
125 g of neutraliser are used to neutralise one L of formalin 4%. Agitate, wait 24 H and the process is completed.