Gull ≥99.9% (metals basis), metalltråd, Ø 0.25 mm (0.01 in)

Leverandør: Thermo Fisher Scientific

00725.G1 00725.G5
00725.G1EA 4250 NOK
00725.G1 00725.G5
Gull ≥99.9% (metals basis), metalltråd, Ø 0.25 mm (0.01 in)
Gull

Purity: 99.9% Physical Form: ≈0.95g/m Odour: Odourless MDL No.: MFCD00003436

Formel: Au
Kokepunkt: 2800 °C (1013 hPa)
Smeltepunkt: 1063 °C
Tetthet: 19,3 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003436
CAS-nummer: 7440-57-5
EINECS: 231-165-9
Merck Indeks: 12,04526

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR