Brom ≥99.8%, liquid

Leverandør: Thermo Fisher Scientific
Danger

00905.14 00905.30 00905.A1 00905.D9
00905.14EA 1050 NOK
00905.14 00905.30 00905.A1 00905.D9
Brom ≥99.8%, liquid
Bromine

Purity: 99.8% CAS No.: 7726-95-6 Physical Form: Liquid Odour: Unpleasant UN No.: 1744 MDL No.: MFCD00010896

Formel: Br₂
Kokepunkt: 58,7 °C (1013 hPa)
Tetthet: 3,12 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010896
CAS-nummer: 7726-95-6
EINECS: 231-778-1
FN: 1744
ADR: 8,I
Merck Indeks: 13,01378

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR