Eosin Y, Certifiable Grade

Leverandør: VWR Chemicals
Warning

Synonymer: Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt

0109-100GEA 1950 NOK
0109-100G
Eosin Y, Certifiable Grade
Eosin Y

Counterstain to hematoxylin. Used for staining exfoliative cytology.

Formel: C₂₀H₆Br₄Na₂O₅
Smeltepunkt: 295…296 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005040
CAS-nummer: 17372-87-1
EINECS: 241-409-6
Merck Indeks: 13,03634

Bestill nå

Spesifikasjon Testresultater

Dye Content >88%
Lambda Max 515 – 518 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR