Sitronsyre monohydrat 99,5-100,5 Anhydrous basis, powder_crystalline USP, FCC, J.T.Baker®

Leverandør: Avantor
Warning

Synonymer: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

0120-05EA 5275 NOK
0120-05
Sitronsyre monohydrat 99,5-100,5 Anhydrous basis, powder_crystalline USP, FCC, J.T.Baker®
Sitronsyre monohydrat
Formel: C₆H₈O₇.H₂O
MW: 210.14 g/mol
Smeltepunkt: 149…151 °C
Tetthet: 1.54 g/cm³
Flash punkt: 173.9 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149972
CAS-nummer: 5949-29-1
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Indeks: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Food GMP Manufactured Product 99.5 - 100.5%
GMP Manufactured Product Passes Test
Meets F.C.C. Requirements Passes Test
Meets U.S.P Requirements Passes Test
Preserve in Tight Containers 7.5 - 9.0%
CAUTION: For Manufacturing, processing or repackaging ≤0.05%
Bulk Food Chemical ≤0.036%
Bulk Pharmaceutical Chemical ≤0.015%
USP - Assay (C₆H₈O₇) (anhydrous basis) 99.5 - 100.5 %
USP - Clarity of Solution Passes Test
USP - Color of Solution Passes Test
USP - Identification (USP) Passes Test
USP - Water (H₂O) (U.S.P.) 7.5 - 9.0 %
USP - Residue on Ignition ≤ 0.05 %
USP - Oxalic Acid (USP) ≤ 0.036 %
USP - Sulfate (SO₄) ≤ 0.015 %
USP - Readily Carbonizable Substances Passes Test
FCC - Assay (C₆H₈O₇) (anhydrous basis) 99.5 - 100.5 %
FCC - Identification (FCC) Passes Test
FCC - Lead (Pb) ≤ 0.5 mg/kg
FCC - Oxalate (C₂O₄) (FCC) ≤100 mg/kg
FCC - Readily Carbonizable Substances Passes Test
FCC - Residue on Ignition ≤ 0.05 %
FCC - Water (H₂O) (F.C.C.) ≤ 8.8 %
Must be subjected to further processing during the preparation of
injectable dosage forms.
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP <232>) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR