Jern(II)sulfat heptahydrat, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Leverandør: Avantor
Warning
0126.0500EA 470 NOK
0126.0500
Jern(II)sulfat heptahydrat, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Jern(II)sulfat heptahydrat
Formel: FeSO₄·7H₂O
Smeltepunkt: 64 °C
Tetthet: 1,9 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149719
CAS-nummer: 7782-63-0
EINECS: 231-753-5
Merck Indeks: 13,04091
Bestill nå
Spesifikasjon Testresultater
Meets ACS Specifications
Assay (by KMnO4 titrn.) min. 99.0 %
Calcium (Ca) max. 0.005 %
Chloride (Cl) max. 0.001 %
Copper (Cu) max. 0.005 %
Potassium (K) max. 0.002 %
Magnesium (Mg) max. 0.002 %
Manganese (Mn) max. 0.05 %
Sodium (Na) max. 0.02 %
Zinc (Zn) max. 0.005 %
Ferric ion (Fe³+) max. 0.1 %
Insoluble Matter max. 0.01 %
Phosphate (PO4) max. 0.001 %