Oksalsyre (Etandisyre) dihydrat, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Warning

0187.1000EA 1225 NOK
0187.1000
Oksalsyre (Etandisyre) dihydrat, BAKER ANALYZED® ACS, J.T.Baker®
Oksalsyre (Etandisyre) dihydrat
Formel: HO₂CCO₂H·2H₂O
Kokepunkt: 149…160 °C (1013 hPa)
Smeltepunkt: 101 °C
Tetthet: 1,65 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149102
CAS-nummer: 6153-56-6
EINECS: 205-634-3
FN: 3261
ADR: 8,III
REACH: 01-2119534576-33
Merck Indeks: 13,06980
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Exceeds ACS Specifications
Assay 99.5 - 102.5 %
Insoluble Matter max. 0.005 %
Nitrogen Compounds (as N) max. 0.001 %
Residue after Ignition max. 0.01 %
Substances Darkened by Hot H2SO4 passes test
Sulfate (SO4) max. 0.002 %
Trace Impurities (in ppm)
Calcium (Ca) max. 5
Chloride (Cl) max. 5
Iron (Fe) max. 2
Heavy Metals (as Pb) max. 5

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR