Organic reference standard, N,N-Dimethylallylamine, analytical standard, Supelco®
Leverandør: Merck
01987-1ML
01987-1MLEA 555 NOK
01987-1ML
Organic reference standard, N,N-Dimethylallylamine, analytical standard, Supelco®
Standarder Organiske standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7ba7d76cdf6dfb28c3bcd40b795cc63fd0d8b22&vid=3fd74f9451da4aab44df97523385d6bf7b60ab29&prid=web&clid=SAPDAM
Bestill nå
SPESIFIKASJONER
Beskrivelse N,N-Dimethylallylamine
CAS-nr 2155-94-4
Formel C₅H₁₁N