Kaliumhydrogenortofosfat, BAKER, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor

0242.1000EA 880 NOK
0242.1000
Kaliumhydrogenortofosfat, BAKER, Laboratory Reagent, J.T.Baker®
Kaliumhydrogenortofosfat
Formel: K₂HPO₄
Smeltepunkt: 340 °C
Tetthet: 2,3 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011383
CAS-nummer: 7758-11-4
EINECS: 231-834-5
Merck Indeks: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay 98.0 - 100.5 %
Carbonate (CO3) passes test
Arsenic (As) max. 3 ppm
Chloride (Cl) max. 0.03 %
Iron (Fe) max. 0.003 %
Sodium (Na) passes test
Heavy Metals (as Pb) max. 0.001 %
Identification passes test
Insoluble Substances max. 0.2 %
Limit of fluoride max. 0.001 %
Loss on Drying at 105°C max. 1.0 %
pH of 5% Solution at 25°C 8.5 - 9.6
Sulfate (SO4) max. 0.1 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR