2-Oktanon, Supelco®

Leverandør: Merck

Synonymer: Octan-2-one

M-Clarity™ quality level = MQ100

02479-5ML
02479-5MLEA 1460 NOK
02479-5ML
2-Oktanon, Supelco®
2-Oktanon
Formel: C₈H₁₆O
Kokepunkt: 173 °C (1013 hPa)
Smeltepunkt: –16 °C
Tetthet: 0,8171 g/cm³ (20 °C)
Flash punkt: 56 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009540
CAS-nummer: 111-13-7
EINECS: 203-837-1
FN: 1993
ADR: 3,III
Merck Indeks: 13,04731

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR