Pyridin, Supelco®

Leverandør: Merck
Danger

Synonymer: Azabenzene, Azine

M-Clarity™ quality level = MQ100

02486-1ML
02486-1MLEA 490 NOK
02486-1ML
Pyridin, Supelco®
Pyridine
Formel: C₅H₅N
Kokepunkt: 115 °C (1013 hPa)
Smeltepunkt: –44 °C
Tetthet: 0,983 g/cm³ (20 °C)
Flash punkt: 17 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011732
CAS-nummer: 110-86-1
EINECS: 203-809-9
FN: 1282
ADR: 3,II
Merck Indeks: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8975fe7b17ff5ccac3461ac47553af20f44756f&vid=0e6ca2ac002c180adb9f123a666d8f8ad7874da0&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR