Kloroform stabilised, Supelco®

Leverandør: Merck
Danger

Synonymer: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

M-Clarity™ quality level = MQ100

02487-5ML
02487-5MLEA 1850 NOK
02487-5ML
Kloroform stabilised, Supelco®
Kloroform
Formel: CHCl₃
MW: 119.38 g/mol
Kokepunkt: 61.7
Smeltepunkt: –63 °C
Tetthet: 1.4832 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000826
CAS-nummer: 67-66-3
EINECS: 200-663-8
FN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Indeks: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bce0c94578071018b29f5b3ea36ad46b2ccb3c9b&vid=b18648d677a146979fa0cce55e4d5a942140206e&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR