Dodecamolybdofosforsyre hydrat solution ready-to-use spray and plunge reagent for chromatography, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Danger

Synonymer: Molybdophosphoric acid hydrate, 12-Molybdophosphoric acid hydrate, Phosphomolybdic acid hydrate

02553-100ML 02553-500ML
02553-100MLEA 362 NOK
02553-100ML 02553-500ML
Dodecamolybdofosforsyre hydrat solution ready-to-use spray and plunge reagent for chromatography, Sigma-Aldrich®
Dodecamolybdofosforsyre hydrat

Phosphomolybdic acid solution,ready-to-use spray and plunge reagent, for chromatography (Molybdophosphoric acid)

Formel: H₃[P(Mo₃O₁₀)₄]·nH₂O
MDL nummer: MFCD00149913
CAS-nummer: 51429-74-4
ADR: 3,II

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR