Rhodamin B solution 0,2% in isopropanol (IPA) derivatisation reagent, for TLC, Supelco®

Leverandør: Merck
Danger

Synonymer: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride

M-Clarity™ quality level = MQ100

02558-100ML 02558-500ML
02558-100MLEA 450 NOK
02558-100ML 02558-500ML
Rhodamin B solution 0,2% in isopropanol (IPA) derivatisation reagent, for TLC, Supelco®
Rhodamin B

Ready-to-use spray and dip reagent for chromatography

Formel: C₂₈H₃₁ClN₂O₃
MDL nummer: MFCD00011931
CAS-nummer: 81-88-9
FN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e2e4b2bb0a483871c494314ff0a4ef766095de2&vid=102246dc474c4665f2a5d41a86ffac401329bc6d&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR