1,2-Diklorpropan, Supelco®

Leverandør: Merck
Danger

Synonymer: Propylene dichloride

M-Clarity™ quality level = MQ100

02577-1ML
02577-1MLEA 290 NOK
02577-1ML
1,2-Diklorpropan, Supelco®
1,2-Diklorpropan
Formel: ClH₂CCH₂CH₂Cl
MW: 112.99 g/mol
Kokepunkt: 96
Smeltepunkt: –100 °C
Tetthet: 1.158 g/cm³ (20 °C)
Flash punkt: 15
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000868
CAS-nummer: 78-87-5
EINECS: 201-152-2
FN: 1279
ADR: 3 II
Merck Indeks: 12,08039
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68aa2c1593fbab47859c4964daf641de70f62897&vid=603b8142bb9de90fe812f56658bc4e5b34b6761c&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR