Ortofosforsyre ≥85.0-87.0%, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Danger


Management of Change (MOC) category = N

0260-08EA 28225 NOK
0260-08 0260-07 0260-15 0260-03 0260-01 0260-33 0260-06
Ortofosforsyre ≥85.0-87.0%, BAKER ANALYZED® ACS, J.T.Baker®
Ortofosforsyre

The quality of BAKER ANALYZED™ ACS Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.

Formel: H₃PO₄
Smeltepunkt: 28 °C
Tetthet: 1,69 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011340
CAS-nummer: 7664-38-2
EINECS: 231-633-2
FN: 1805
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements 85.0 - 87.0%
For Laboratory,Research,or Manufacturing Use ≤0.002%
Assay (H₃PO₄) (by acidimetry) 85.0 - 87.0 %
Calcium (Ca) ≤ 0.002 %
Color (APHA) ≤ 10
Insoluble Matter ≤ 0.001 %
ACS - Magnesium (Mg) ≤ 0.002 %
Sulfate (SO₄) ≤ 12 ppm
Volatile Acids (as CH₃COOH) ≤ 0.001 %
Reducing Substances Passes Test
Chloride (Cl) ≤ 3 ppm
Nitrate (NO₃) ≤ 5 ppm
Trace Impurities - Antimony (Sb) ≤ 20.000 ppm
Trace Impurities - Arsenic (As) ≤ 0.500 ppm
Trace Impurities - Iron (Fe) ≤ 10 ppm
Heavy Metals (as Pb) ≤ 8 ppm
Trace Impurities - Manganese (Mn) ≤ 0.500 ppm
Trace Impurities - Potassium (K) ≤ 40.000 ppm
Trace Impurities - Sodium (Na) ≤ 200.000 ppm
Exceeds A.C.S. Specifications
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR