Tetramethylammonium sulphate ≥99.0% (by titrimetric analysis), powder, LiChropur™ for ion pair chromatography, Supelco®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ100

02799-10G
02799-10GEA 4420 NOK
02799-10G
Tetramethylammonium sulphate ≥99.0% (by titrimetric analysis), powder, LiChropur™ for ion pair chromatography, Supelco®
Tetramethylammonium sulphate
Formel: C₈H₂₄N₂O₄S
MDL nummer: MFCD00012139
CAS-nummer: 14190-16-0
EINECS: 238-043-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da619d9062b48074680823498a7312c7d9a19989&vid=da619d9062b48074680823498a7312c7d9a19989&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Color) Colorless or White
Titration (Ion Exchange) 99.0 - 101.0 %
Suitability Test (HPLC) RP-Gradient Test Corresponds to Standard
Proton NMR Spectrum Conforms to Structure
Test By UV Spectroscopy 500nm≤ 0.02
Cyclic Voltammogram Corresponds to Standard CV
Residue (Filter Test) No Residue
Filtertest (Method) 3g in 30ml Water (10%)
Recommended Retest Period 42 Months

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR