ACES ≥99%, high purity

Leverandør: VWR Chemicals

Synonymer: ACES, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid

0285-100GEA 3150 NOK
0285-100G
ACES ≥99%, high purity
ACES

Low interference with biological processes and biological membranes (penetration, solubilisation, adsorption on surface).


  • Water soluble
  • Non-toxic
  • Low UV absorption at wavelengths less than 260 nm

Formel: H₂NCOCH₂NHCH₂CH₂SO₃H
MW: 182.2 g/mol
Smeltepunkt: 270…295 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008030
CAS-nummer: 7365-82-4
EINECS: 230-908-4
Merck Indeks: 13,00037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d8d301fc0de89d24309fbfdbaedb924c1debb4a&vid=a5fb2045198b642d124372f31d901db2e2d25597&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Abs.@280 nm (5%, Water) ≤0.02
pH (1%, Water) @25 °C 3.6 – 4.4
pKa @25 °C 6.58 – 6.98
Purity ≥99%
Water (Karl Fisher) ≤1%

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR