Magnesiumklorid heksahydrat 99-102% ACS

Leverandør: VWR Chemicals

0288-1KGEA 2775 NOK
0288-1KG 0288-100G 0288-500G
Magnesiumklorid heksahydrat 99-102% ACS
Magnesiumklorid heksahydrat
Formel: MgCl₂·6H₂O
Kokepunkt: 1412 °C (1013 hPa)
Smeltepunkt: 117 °C
Tetthet: 1,57 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149781
CAS-nummer: 7791-18-6
EINECS: 232-094-6
Merck Indeks: 13,05684

Bestill nå

Spesifikasjon Testresultater

Ammonium <0.002%
Barium ≤0.005%
Calcium ≤0.01%
Heavy Metals (as Pb) ≤0.0005%
Insolubles ≤0.005%
Iron ≤0.0005%
Manganese ≤0.0005%
Nitrate ≤0.001%
Phosphate ≤0.0005%
Potassium ≤0.005%
Purity 99 – 102%
Sodium ≤0.005%
Strontium ≤0.005%
Sulphate ≤0.002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR