Natriumjodid, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor
Warning

0293.5000EA 12150 NOK
0293.5000
Natriumjodid, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Natriumjodid
Formel: NaI
Kokepunkt: 1300 °C (1013 hPa)
Smeltepunkt: 662 °C
Tetthet: 3,67 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003532
CAS-nummer: 7681-82-5
EINECS: 231-679-3
FN: 3077
ADR: 9,III
Merck Indeks: 13,08705

Bestill nå

Spesifikasjon Testresultater

Assay (dried basis) min. 99.0 %
Barium (Ba) max. 0.002 %
Iron (Fe) max. 5 ppm
Heavy Metals (as Pb) max. 5 ppm
Insoluble Matter max. 0.005 %
Iodate (IO3) max. 4 ppm
Nitrogen Compounds (as N) max. 0.001 %
pH of 5% Solution at 25°C 7.5 - 9.5
Phosphate (PO4) max. 0.002 %
Sulfate (SO4) max. 150 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR