Kobber(II)sulfat pentahydrat 98-102% ACS

Leverandør: VWR Chemicals
Warning

0330-2.5KGEA 4700 NOK
0330-2.5KG 0330-500G
Kobber(II)sulfat pentahydrat 98-102% ACS
Kobber(II)sulfat pentahydrat
Formel: CuSO₄·5H₂O
Tetthet: 2,284 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149681
CAS-nummer: 7758-99-8
EINECS: 231-847-6
FN: 3077
ADR: 9,III
REACH: 01-2119520566-40
Merck Indeks: 13,02682

Bestill nå

Spesifikasjon Testresultater

Calcium <0.005%
Chloride ≤0.001%
Insolubles ≤0.005%
Iron ≤0.003%
Nickel ≤0.005%
Nitrogen Compounds (as N) ≤0.002%
Potassium ≤0.01%
Purity 98% – 102%
Sodium ≤0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR