L(+)-Lactic acid 85-90% ACS

Leverandør: VWR Chemicals
Danger

Synonymer: (S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid

0349-4LEA 18575 NOK
0349-4L
L(+)-Lactic acid 85-90% ACS
L(+)-Lactic acid
Formel: C₃H₆O₃
MW: 90,08 g/mol
Kokepunkt: 122 °C (20 hPa)
Smeltepunkt: 16,8 °C
Tetthet: 1,357 g/cm³ (20 °C)
Flash punkt: 113 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064266
CAS-nummer: 79-33-4
EINECS: 209-954-4
Merck Indeks: 13,05351

Bestill nå

Spesifikasjon Testresultater

Chloride <0.001%
Heavy Metals (as Pb) ≤0.0005%
Iron ≤0.0005%
Purity 85 – 90%
Residue on Ignition ≤0.02%
Substances Darkened by H2SO4 Pass
Sulphate ≤0.002%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR