MALDI validation set polystyrene Mp ~700 - 70000, certified reference material, Supelco®
Leverandør: Merck
03565-1EA
03565-1EAEA 21660 NOK
03565-1EA
MALDI validation set polystyrene Mp ~700 - 70000, certified reference material, Supelco®
Standarder kromatografi standarder
MALDI Standards are suitable for the validation and comparison of MALDI-ToF instruments. Certificates provide MALDI spectra including Mw, Mn and Mp values of select narrow polymers, which differ in molar mass.
This set inclces six different polystyrene standards ranging from Mp ~700 to ~70000.
Each standard is characterized with GPC, a second absolute method (light scattering or H-NMR) and MALDI-ToF-MS.