EDTA dinatrium salt dihydrat ≥99.0% for HPLC, Fluka™

Leverandør: Honeywell Chemicals
Warning

03682-10G 03682-50G
03682-10GEA 890 NOK
03682-10G 03682-50G
EDTA dinatrium salt dihydrat ≥99.0% for HPLC, Fluka™
EDTA dinatrium salt dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Smeltepunkt: 244-250 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150037
CAS-nummer: 6381-92-6
EINECS: 230-944-0
Merck Indeks: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay 99.0 - 101.0 %
Suitability test (HPLC) complying
Colour of the solution complying
Clearness of the solution complying
Absorbance at 500 nm Max. 0.02
Absorbance at 290 nm Max. 0.02
Absorbance at 280 nm Max. 0.03
Absorbance at 270 nm Max. 0.05
Absorbance at 260 nm Max. 0.15
Aluminium (Al) Max. 5 ppm
Barium (Ba) Max. 5 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Molybdenum (Mo) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Total S (as SO₄) Max. 50 ppm
Chloride (Cl) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR