Sodium-3-[ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulphonate hydrate ≥98.0% (dried basis, T) for spectrophotometric determination H₂O₂, Supelco®

Leverandør: Merck

Synonymer: 3-[Ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulphonic acid Sodium Salt, TOOS, 3-(N-Ethyl-3-methylanilino)-2-hydroxypropanesulphonic acid sodium salt

M-Clarity™ quality level = MQ100

04340-500MG
04340-500MGEA 1210 NOK
04340-500MG
Sodium-3-[ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulphonate hydrate ≥98.0% (dried basis, T) for spectrophotometric determination H₂O₂, Supelco®
Sodium-3-[ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulphonate hydrate

Water-soluble reagent for the enzymatic photometric determination of hydrogen peroxide.

Formel: C₁₂H₁₉NO₄S
MW: 295.33 g/mol
MDL nummer: MFCD00040641
CAS-nummer: 82692-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9cdb1b944164c7828f7fe6456ab4b3ff9ff4b3b&vid=f7ba75497108facc85808c9439c2dc3f2f4c640a&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR