Organic reference standard, Bisphenol E, analytical standard, Supelco®
Leverandør: Merck
04487-100MG
04487-100MGEA 545 NOK
04487-100MG
Organic reference standard, Bisphenol E, analytical standard, Supelco®
Standarder Organiske standarder
Bisphenol E may be used as a reference standard in the determination of bisphenol E in beverages using on-line solid-phase extraction with fast liquid chromatography coupled with tandem mass spectrometry (on-line SPE fast LC-MS/MS).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82851d90a86da4dd53f1f4f849fac967dd3217ee&vid=82851d90a86da4dd53f1f4f849fac967dd3217ee&prid=web&clid=SAPDAM