VWR® ARISTAR®, Multi-Element Quality Control Standard 1 mg/L, 28 Elements in Nitric Acid 5% for ICP
Leverandør: VWR Chemicals
05200.185EA 1095 NOK
05200.185
VWR® ARISTAR®, Multi-Element Quality Control Standard 1 mg/L, 28 Elements in Nitric Acid 5% for ICP
Standarder Kationstandarder Multi-Element Standards
28 elements 1 mg/L: Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.