Supelco® PESTANAL® Organic Reference Standard, Propoxur-(Isopropoxy-D₇)

Leverandør: Merck

05351-5MG
05351-5MGEA 1950 NOK
05351-5MG
Supelco® PESTANAL® Organic Reference Standard, Propoxur-(Isopropoxy-D₇)
Standarder Organiske standarder

  • GHS signal: GHS06, GHS09


NB: P273 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P310

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cad98065be3cca0ca0bb4115e8fc4f639e40e9e&vid=f0988bfff4de46c85cd70bae7d74ea55d1f7fc00&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) 2-Isopropoxy-d₇-phenyl N-methylcarbamate; Propoxur-d₇
M-Clarity quality level MQ100
CAS-nr 114-26-1
Formel C₁₁D₇H₈NO₃
Molekylvekt 216,28
Hardhetsklasse beskrivelse H300 + H330 - H311 - H410
Flammepunkt Not applicable
Fraktforhold STD
Lagringsforhold 2...8 °C
Lagringstemp 2...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR