Aluminiumoksid neutral, pulver, BAKER ANALYZED®, J.T.Baker®

Leverandør: Avantor
Danger


Management of Change (MOC) category = N

0540-01EA 3150 NOK
0540-01
Aluminiumoksid neutral, pulver, BAKER ANALYZED®, J.T.Baker®
Aluminium oxide neutral
Formel: Al₂O₃
Kokepunkt: 2980 °C (1013 hPa)
Smeltepunkt: 2045 °C
Tetthet: 3,94 g/cm³ (20 °C)
MDL nummer: MFCD00003424
CAS-nummer: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Indeks: 13,00355

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use 7.0 - 7.8
pH of 5% Slurry at 20°C 6.0 - 7.5
Adsorption, mg o-nitroaniline/g ≥1.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR