Natriummolybdat(VI) dihydrat ≥99.5% ACS

Leverandør: VWR Chemicals

Synonymer: Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate

0541-250GEA 6450 NOK
0541-250G
Natriummolybdat(VI) dihydrat ≥99.5% ACS
Natriummolybdat(VI) dihydrat
Formel: Na₂MoO₄·2H₂O
Smeltepunkt: 687 °C
Tetthet: 3,28 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149170
CAS-nummer: 10102-40-6
EINECS: 231-551-7
Merck Indeks: 13,08718

Bestill nå

Spesifikasjon Testresultater

Ammonium <0.001%
Chloride ≤0.005%
Heavy Metals (as Pb) ≤0.0005%
Insolubles ≤0.005%
Iron ≤0.001%
pH (5%, Water) @25 °C 7 – 10.5
Phosphate ≤0.0005%
Purity 99.5 – 103%
Sulphate ≤0.015%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR