Kalsiumklorid dihydrat 99-105% ACS

Leverandør: VWR Chemicals
Warning

0556-1KGEA 3625 NOK
0556-1KG 0556-2.5KG 0556-500G
Kalsiumklorid dihydrat 99-105% ACS
Kalsiumklorid dihydrat
Formel: CaCl₂·2H₂O
Kokepunkt: 1600 °C (1013 hPa)
Smeltepunkt: 176 °C
Tetthet: 1,85 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149613
CAS-nummer: 10035-04-8
EINECS: 233-140-8
Merck Indeks: 13,01660

Bestill nå

Spesifikasjon Testresultater

Ammonium <0.005%
Assay 99 – 105%
Barium ≤0.005%
Heavy Metals (as Pb) ≤0.0005%
Insolubles ≤0.01%
Iron ≤0.001%
Magnesium ≤0.005%
Oxidizers (as NO₃) ≤0.003%
pH (5%, Water) @25 °C 4.5 – 8.5
Potassium ≤0.01%
Sodium ≤0.02%
Strontium ≤0.1%
Sulphate ≤0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR