Diammoniumhydrogenortofosfat 98 ACS

Leverandør: VWR Chemicals

Synonymer: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

0569-1KGEA 4125 NOK
0569-1KG 0569-2.5KG 0569-500G
Diammoniumhydrogenortofosfat 98 ACS
Diammoniumhydrogenortofosfat
Formel: (NH₄)₂HPO₄
MW: 132.06 g/mol
Smeltepunkt: 185
Tetthet: 1.619 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010891
CAS-nummer: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Indeks: 13,00546

Bestill nå

Spesifikasjon Testresultater

Calcium <0.001%
Chloride ≤0.001%
Heavy Metals (as Pb) ≤0.001%
Insolubles ≤0.005%
Iron ≤0.001%
Magnesium ≤0.0005%
Nitrate ≤0.003%
pH (5%, Water) @25 °C 7.7 – 8.1
Potassium ≤0.005%
Purity ≥98%
Sodium ≤0.005%
Sulphate ≤0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR