Safranine O, Certifiable Grade

Leverandør: VWR Chemicals

Synonymer: CI 50240

0574-100GEA 10500 NOK
0574-100G 0574-25G
Safranine O, Certifiable Grade
Safranine O

Part Fleming Triple stain, a chromosome stain prepared with light green SF and in Gram stain.

Formel: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
Tetthet: 1.447 g/cm³ (22 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011759
CAS-nummer: 477-73-6
EINECS: 207-518-8

Bestill nå

Spesifikasjon Testresultater

Absorbance maximum 527 – 533 nm
Dye Content (Anhydrous) ≥80%
Loss on Drying ≤4%
P-15/P+15 1.1 – 1.32


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR